Anti-IL1B Rat Monoclonal Antibody [clone: 424823]

Supplier: R & D Systems
MAB3340
220020-402EA 561.11 USD
220020-402
Anti-IL1B Rat Monoclonal Antibody [clone: 424823]
Antibodies
Type: Primary
Antigen: IL1B
Clonality: Monoclonal
Clone: 424823
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Horse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR