Anti-PTGDR2 Mouse Monoclonal Antibody [clone: 301109]

Supplier: R & D Systems
MAB33381
102885-570EA 577.02 USD
102885-570
Anti-PTGDR2 Mouse Monoclonal Antibody [clone: 301109]
Antibodies
Type: Primary
Antigen: PTGDR2
Clonality: Monoclonal
Clone: 301109
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR