Anti-BMI1 Mouse Monoclonal Antibody [clone: 384515]

Supplier: R & D Systems
MAB33341
220020-398EA 471.45 USD
220020-398
Anti-BMI1 Mouse Monoclonal Antibody [clone: 384515]
Antibodies
Type: Primary
Antigen: BMI1
Clonality: Monoclonal
Clone: 384515
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR