Anti-CXCL6 Mouse Monoclonal Antibody [clone: 60910]

Supplier: R & D Systems
MAB333
220020-397EA 772.25 USD
220020-397
Anti-CXCL6 Mouse Monoclonal Antibody [clone: 60910]
Antibodies
Type: Primary
Antigen: CXCL6
Clonality: Monoclonal
Clone: 60910
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR