Anti-LAG3 Rat Monoclonal Antibody [clone: 631501]

Supplier: R & D Systems
MAB33281
102885-568EA 633.42 USD
102885-568
Anti-LAG3 Rat Monoclonal Antibody [clone: 631501]
Antibodies
Type: Primary
Antigen: LAG3
Clonality: Monoclonal
Clone: 631501
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR