Anti-CD48 Rat Monoclonal Antibody [clone: 331504]

Supplier: R & D Systems
MAB3327
220020-396EA 594.37 USD
220020-396
Anti-CD48 Rat Monoclonal Antibody [clone: 331504]
Antibodies
Type: Primary
Antigen: CD48
Clonality: Monoclonal
Clone: 331504
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR