Anti-S100 G Mouse Monoclonal Antibody [clone: 401025]

Supplier: R & D Systems
MAB3320
220020-394EA 633.42 USD
220020-394
Anti-S100 G Mouse Monoclonal Antibody [clone: 401025]
Antibodies
Simple Western

Type: Primary
Antigen: S100 G
Clonality: Monoclonal
Clone: 401025
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR