Anti-NEUROG2 Mouse Monoclonal Antibody [clone: 7G4]

Supplier: R & D Systems
MAB3314
220020-391EA 623.29 USD
220020-391
Anti-NEUROG2 Mouse Monoclonal Antibody [clone: 7G4]
Antibodies
Type: Primary
Antigen Symbol: NEUROG2
Antigen Name: neurogenin 2
Antigen Synonyms: Ngn2
Clonality: Monoclonal
Clone: 7G4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR