Anti-CXCR2 Mouse Monoclonal Antibody [clone: 48311]

Supplier: R & D Systems
MAB331-100
102885-566EA 655.11 USD
102885-566
Anti-CXCR2 Mouse Monoclonal Antibody [clone: 48311]
Antibodies
Type: Primary
Antigen: CXCR2
Clonality: Monoclonal
Clone: 48311
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR