Anti-CFI Mouse Monoclonal Antibody [clone: 271203]

Supplier: R & D Systems
MAB3307
220020-387EA 594.37 USD
220020-387
Anti-CFI Mouse Monoclonal Antibody [clone: 271203]
Antibodies
Type: Primary
Antigen: CFI
Clonality: Monoclonal
Clone: 271203
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR