Anti-F7 Rat Monoclonal Antibody [clone: 406722]

Supplier: R & D Systems
MAB3305
220020-386EA 561.11 USD
220020-386
Anti-F7 Rat Monoclonal Antibody [clone: 406722]
Antibodies
Type: Primary
Antigen Symbol: F7
Antigen Name: coagulation factor VII
Antigen Synonyms: AI132620, Cf7, FVII
Clonality: Monoclonal
Clone: 406722
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR