Anti-MGMT Rat Monoclonal Antibody [clone: 300008]

Supplier: R & D Systems
MAB3299
220020-383EA 577.02 USD
220020-383
Anti-MGMT Rat Monoclonal Antibody [clone: 300008]
Antibodies
Type: Primary
Antigen: MGMT
Clonality: Monoclonal
Clone: 300008
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR