Anti-HSPE1 Mouse Monoclonal Antibody [clone: 330629]

Supplier: R & D Systems
MAB3298
220020-382EA 594.37 USD
220020-382
Anti-HSPE1 Mouse Monoclonal Antibody [clone: 330629]
Antibodies
Type: Primary
Antigen: HSPE1
Clonality: Monoclonal
Clone: 330629
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR