Anti-SMC1A Mouse Monoclonal Antibody [clone: 317705]

Supplier: R & D Systems
MAB3294
220020-381EA 561.11 USD
220020-381
Anti-SMC1A Mouse Monoclonal Antibody [clone: 317705]
Antibodies
Type: Primary
Antigen: SMC1A
Clonality: Monoclonal
Clone: 317705
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR