Anti-CSF1R Mouse Monoclonal Antibody [clone: 61708]

Supplier: R & D Systems
MAB329
220020-378EA 772.25 USD
220020-378
Anti-CSF1R Mouse Monoclonal Antibody [clone: 61708]
Antibodies
Type: Primary
Antigen: CSF1R
Clonality: Monoclonal
Clone: 61708
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR