Anti-PLCG1 Rat Monoclonal Antibody [clone: 479819]

Supplier: R & D Systems
MAB32881
220020-377EA 561.11 USD
220020-377
Anti-PLCG1 Rat Monoclonal Antibody [clone: 479819]
Antibodies
Type: Primary
Antigen: PLCG1
Clonality: Monoclonal
Clone: 479819
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR