Anti-PLCG1 Mouse Monoclonal Antibody [clone: 346130]

Supplier: R & D Systems
MAB3288
220020-376EA 594.37 USD
220020-376
Anti-PLCG1 Mouse Monoclonal Antibody [clone: 346130]
Antibodies
Type: Primary
Antigen: PLCG1
Clonality: Monoclonal
Clone: 346130
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR