Anti-HSP90AB1 Mouse Monoclonal Antibody [clone: 341346]

Supplier: R & D Systems
MAB32861
220020-375EA 577.02 USD
220020-375
Anti-HSP90AB1 Mouse Monoclonal Antibody [clone: 341346]
Antibodies
Type: Primary
Antigen: HSP90AB1
Clonality: Monoclonal
Clone: 341346
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR