Anti-HSP90 Mouse Monoclonal Antibody [clone: 341320]

Supplier: R & D Systems
MAB3286
220020-374EA 600.15 USD
220020-374
Anti-HSP90 Mouse Monoclonal Antibody [clone: 341320]
Antibodies
Simple Western

Type: Primary
Antigen: HSP90
Clonality: Monoclonal
Clone: 341320
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR