Anti-CCL16 Mouse Monoclonal Antibody [clone: 70218]

Supplier: R & D Systems
MAB328
220020-373EA 763.57 USD
220020-373
Anti-CCL16 Mouse Monoclonal Antibody [clone: 70218]
Antibodies
Type: Primary
Antigen: CCL16
Clonality: Monoclonal
Clone: 70218
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR