Anti-TRAF2 Mouse Monoclonal Antibody [clone: 324522]

Supplier: R & D Systems
MAB3277
220020-370EA 594.37 USD
220020-370
Anti-TRAF2 Mouse Monoclonal Antibody [clone: 324522]
Antibodies
Type: Primary
Antigen: TRAF2
Clonality: Monoclonal
Clone: 324522
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR