Anti-MAPK11 Mouse Monoclonal Antibody [clone: 280071]

Supplier: R & D Systems
MAB3274
220020-369EA 577.02 USD
220020-369
Anti-MAPK11 Mouse Monoclonal Antibody [clone: 280071]
Antibodies
Type: Primary
Antigen: MAPK11
Clonality: Monoclonal
Clone: 280071
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR