Anti-RALA Rat Monoclonal Antibody [clone: 399527]

Supplier: R & D Systems
MAB3270
220020-368EA 561.11 USD
220020-368
Anti-RALA Rat Monoclonal Antibody [clone: 399527]
Antibodies
Type: Primary
Antigen: RALA
Clonality: Monoclonal
Clone: 399527
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR