Anti-CCL13 Mouse Monoclonal Antibody [clone: 73506]

Supplier: R & D Systems
MAB327
220020-367EA 772.25 USD
220020-367
Anti-CCL13 Mouse Monoclonal Antibody [clone: 73506]
Antibodies
Type: Primary
Antigen: CCL13
Clonality: Monoclonal
Clone: 73506
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR