Anti-RSPO2 Rat Monoclonal Antibody [clone: 879709]

Supplier: R & D Systems
MAB3266
102885-562EA 594.37 USD
102885-562
Anti-RSPO2 Rat Monoclonal Antibody [clone: 879709]
Antibodies
Type: Primary
Antigen: RSPO2
Clonality: Monoclonal
Clone: 879709
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR