Anti-MSLN Mouse Monoclonal Antibody [clone: 618923]

Supplier: R & D Systems
MAB32653
220020-366EA 644.98 USD
220020-366
Anti-MSLN Mouse Monoclonal Antibody [clone: 618923]
Antibodies
Simple Western

Type: Primary
Antigen: MSLN
Clonality: Monoclonal
Clone: 618923
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR