Anti-MSLN Mouse Monoclonal Antibody [clone: 346633]

Supplier: R & D Systems
MAB3265
220020-363EA 608.83 USD
220020-363
Anti-MSLN Mouse Monoclonal Antibody [clone: 346633]
Antibodies
Type: Primary
Antigen: MSLN
Clonality: Monoclonal
Clone: 346633
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR