Anti-SIGLEC11 Mouse Monoclonal Antibody [clone: 705904]

Supplier: R & D Systems
MAB3258
102885-560EA 600.15 USD
102885-560
Anti-SIGLEC11 Mouse Monoclonal Antibody [clone: 705904]
Antibodies
Type: Primary
Antigen: SIGLEC11
Clonality: Monoclonal
Clone: 705904
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR