Anti-RLN1 Rat Monoclonal Antibody [clone: 361513]

Supplier: R & D Systems
MAB32572
220020-361EA 763.57 USD
220020-361
Anti-RLN1 Rat Monoclonal Antibody [clone: 361513]
Antibodies
Type: Primary
Antigen: RLN1
Clonality: Monoclonal
Clone: 361513
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR