Anti-RLN1 Mouse Monoclonal Antibody [clone: 351713]

Supplier: R & D Systems
MAB3257
220020-359EA 577.02 USD
220020-359
Anti-RLN1 Mouse Monoclonal Antibody [clone: 351713]
Antibodies
Type: Primary
Antigen: RLN1
Clonality: Monoclonal
Clone: 351713
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR