Anti-MCAM Mouse Monoclonal Antibody [clone: 404722]

Supplier: R & D Systems
MAB3250
220020-358EA 566.89 USD
220020-358
Anti-MCAM Mouse Monoclonal Antibody [clone: 404722]
Antibodies
Type: Primary
Antigen: MCAM
Clonality: Monoclonal
Clone: 404722
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR