Anti-HNE Mouse Monoclonal Antibody [clone: 198960]

Supplier: R & D Systems
MAB3249
220020-356EA 647.88 USD
220020-356
Anti-HNE Mouse Monoclonal Antibody [clone: 198960]
Antibodies
Simple Western

Type: Primary
Antigen: HNE
Clonality: Monoclonal
Clone: 198960
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Multiple
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR