Anti-CML Mouse Monoclonal Antibody [clone: 318003]

Supplier: R & D Systems
MAB3247
220020-354EA 655.11 USD
220020-354
Anti-CML Mouse Monoclonal Antibody [clone: 318003]
Antibodies
Type: Primary
Antigen: CML
Clonality: Monoclonal
Clone: 318003
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR