Anti-MMP8 Mouse Monoclonal Antibody [clone: 400102]

Supplier: R & D Systems
MAB3245
220020-353EA 584.25 USD
220020-353
Anti-MMP8 Mouse Monoclonal Antibody [clone: 400102]
Antibodies
Type: Primary
Antigen: MMP8
Clonality: Monoclonal
Clone: 400102
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR