Anti-CCL14 Rat Monoclonal Antibody [clone: 256409]

Supplier: R & D Systems
MAB3241
220020-352EA 772.25 USD
220020-352
Anti-CCL14 Rat Monoclonal Antibody [clone: 256409]
Antibodies
Type: Primary
Antigen: CCL14
Clonality: Monoclonal
Clone: 256409
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR