Anti-CCL14 Rat Monoclonal Antibody [clone: 256413]

Supplier: R & D Systems
MAB324
220020-351EA 763.57 USD
220020-351
Anti-CCL14 Rat Monoclonal Antibody [clone: 256413]
Antibodies
Type: Primary
Antigen: CCL14
Clonality: Monoclonal
Clone: 256413
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR