Anti-SEMA3E Mouse Monoclonal Antibody [clone: 400513]

Supplier: R & D Systems
MAB32391
220020-350EA 577.02 USD
220020-350
Anti-SEMA3E Mouse Monoclonal Antibody [clone: 400513]
Antibodies
Type: Primary
Antigen: SEMA3E
Clonality: Monoclonal
Clone: 400513
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR