Anti-SEMA3E Mouse Monoclonal Antibody [clone: 400520]

Supplier: R & D Systems
MAB3239
220020-349EA 577.02 USD
220020-349
Anti-SEMA3E Mouse Monoclonal Antibody [clone: 400520]
Antibodies
Type: Primary
Antigen: SEMA3E
Clonality: Monoclonal
Clone: 400520
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR