Anti-SEMA3F Rat Monoclonal Antibody [clone: 741307]

Supplier: R & D Systems
MAB3237
102885-558EA 577.02 USD
102885-558
Anti-SEMA3F Rat Monoclonal Antibody [clone: 741307]
Antibodies
Type: Primary
Antigen: SEMA3F
Clonality: Monoclonal
Clone: 741307
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR