Anti-NFASC Mouse Monoclonal Antibody [clone: 727030]

Supplier: R & D Systems
MAB3235
102885-556EA 594.37 USD
102885-556
Anti-NFASC Mouse Monoclonal Antibody [clone: 727030]
Antibodies
Type: Primary
Antigen Symbol: NFASC
Antigen Name: neurofascin
Antigen Synonyms: NF, NRCAML
Clonality: Monoclonal
Clone: 727030
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR