Anti-MATN2 Rat Monoclonal Antibody [clone: 388207]

Supplier: R & D Systems
MAB3234
220020-348EA 561.11 USD
220020-348
Anti-MATN2 Rat Monoclonal Antibody [clone: 388207]
Antibodies
Type: Primary
Antigen: MATN2
Clonality: Monoclonal
Clone: 388207
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR