Anti-BMP2 Mouse Monoclonal Antibody [clone: 369505]

Supplier: R & D Systems
MAB3229
220020-346EA 566.89 USD
220020-346
Anti-BMP2 Mouse Monoclonal Antibody [clone: 369505]
Antibodies
Type: Primary
Antigen: BMP2
Clonality: Monoclonal
Clone: 369505
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR