Anti-EIF4E Mouse Monoclonal Antibody [clone: 299910]

Supplier: R & D Systems
MAB3228
220020-345EA 594.37 USD
220020-345
Anti-EIF4E Mouse Monoclonal Antibody [clone: 299910]
Antibodies
Type: Primary
Antigen: EIF4E
Clonality: Monoclonal
Clone: 299910
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR