Anti-MEP1A Mouse Monoclonal Antibody [clone: 364312]

Supplier: R & D Systems
MAB3220
220020-344EA 594.37 USD
220020-344
Anti-MEP1A Mouse Monoclonal Antibody [clone: 364312]
Antibodies
Type: Primary
Antigen: MEP1A
Clonality: Monoclonal
Clone: 364312
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR