Anti-PRL Mouse Monoclonal Antibody [clone: 334407]

Supplier: R & D Systems
MAB32191
220020-342EA 594.37 USD
220020-342
Anti-PRL Mouse Monoclonal Antibody [clone: 334407]
Antibodies
Type: Primary
Antigen: PRL
Clonality: Monoclonal
Clone: 334407
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR