Anti-PTP4A3 Mouse Monoclonal Antibody [clone: 334402]

Supplier: R & D Systems
MAB3219
220020-341EA 600.15 USD
220020-341
Anti-PTP4A3 Mouse Monoclonal Antibody [clone: 334402]
Antibodies
Type: Primary
Antigen: PTP4A3
Clonality: Monoclonal
Clone: 334402
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR