Anti-NODAL Mouse Monoclonal Antibody [clone: 784410]

Supplier: R & D Systems
MAB3218
102885-554EA 594.37 USD
102885-554
Anti-NODAL Mouse Monoclonal Antibody [clone: 784410]
Antibodies
Type: Primary
Antigen: NODAL
Clonality: Monoclonal
Clone: 784410
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR