Anti-EGF Mouse Monoclonal Antibody [clone: 420610]

Supplier: R & D Systems
MAB3214
220020-340EA 763.57 USD
220020-340
Anti-EGF Mouse Monoclonal Antibody [clone: 420610]
Antibodies
Type: Primary
Antigen: EGF
Clonality: Monoclonal
Clone: 420610
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR