Anti-FGR Mouse Monoclonal Antibody [clone: 349331]

Supplier: R & D Systems
MAB32071
220020-336EA 561.11 USD
220020-336
Anti-FGR Mouse Monoclonal Antibody [clone: 349331]
Antibodies
Type: Primary
Antigen Symbol: FGR
Antigen Name: FGR proto-oncogene, Src family tyrosine kinase
Antigen Synonyms: SRC2, c-fgr, c-src2, p55-Fgr, p55c-fgr, p58-Fgr, p58c-fgr
Clonality: Monoclonal
Clone: 349331
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR