Anti-FGR Mouse Monoclonal Antibody [clone: 349316]

Supplier: R & D Systems
MAB3207
220020-335EA 594.37 USD
220020-335
Anti-FGR Mouse Monoclonal Antibody [clone: 349316]
Antibodies
Type: Primary
Antigen Symbol: FGR
Antigen Name: Gardner-Rasheed feline sarcoma viral (Fgr) oncogene homolog
Antigen Synonyms: -
Clonality: Monoclonal
Clone: 349316
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR